Assay Details
Gene Symbol: Chrna7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012832.3 | NP_036964.3 | 1 - 2 | 57 | 72 | |
XM_008759498.1 | XP_008757720.1 | 1 - 2 | 117 | 72 | ||
GenBank mRNA | AY574256.1 | - | 1 - 2 | 57 | 72 | |
AY671974.1 | - | 1 - 2 | 60 | 72 | ||
L31619.1 | - | 1 - 2 | 79 | 72 | ||
S53987.1 | - | 98 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
