Assay Details
Gene Symbol: Fnta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012847.1 | NP_036979.1 | 4 - 5 | 563 | 77 | |
GenBank mRNA | BC085758.1 | - | 4 - 5 | 513 | 77 | |
M81225.1 | - | 4 - 5 | 563 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
