Assay Details
Gene Symbol: P4hb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012998.1 | NP_037130.1 | 4 - 5 | 675 | 148 | |
GenBank mRNA | BC061857.1 | - | 4 - 5 | 689 | 148 | |
M21018.1 | - | 4 - 5 | 553 | 148 | ||
M21476.1 | - | 2 - 3 | 252 | 148 | ||
X02918.1 | - | 4 - 5 | 675 | 148 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
