Assay Details
Gene Symbol: Acadsb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013084.2 | NP_037216.1 | 1 - 2 | 54 | 68 | |
XM_008759865.1 | XP_008758087.1 | 1 - 2 | 139 | 68 | ||
XM_008759866.1 | XP_008758088.1 | 1 - 2 | 139 | 68 | ||
XM_008759867.1 | XP_008758089.1 | 1 - 2 | 140 | 68 | ||
GenBank mRNA | FQ210776.1 | - | 1 - 2 | 63 | 68 | |
U64451.1 | - | 1 - 2 | 55 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
