Assay Details
Gene Symbol: Eif2s1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019356.1 | NP_062229.1 | 4 - 5 | 496 | 120 | |
GenBank mRNA | BC087019.1 | - | 4 - 5 | 553 | 120 | |
J02646.1 | - | 4 - 5 | 496 | 120 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
