Assay Details
Gene Symbol: Kcnab3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031652.1 | NP_113840.1 | 10 - 11 | 1223 | 112 | |
XM_008767862.1 | XP_008766084.1 | 10 - 11 | 1261 | 112 | ||
GenBank mRNA | X76723.1 | - | 10 - 11 | 1223 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
