Assay Details
Gene Symbol: Ceacam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033860.1 | NP_001029032.1 | 7 - 8 | 1468 | 66 | |
NM_031755.2 | NP_113943.1 | 6 - 7 | 1341 | 66 | ||
GenBank mRNA | AJ277104.1 | - | 6 - 7 | 1303 | 66 | |
BC061740.1 | - | 7 - 8 | 1454 | 66 | ||
J04963.1 | - | 7 - 8 | 1475 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
