Assay Details
Gene Symbol: Gosr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053584.2 | NP_446036.1 | 1 - 2 | 36 | 118 | |
XM_006246914.1 | XP_006246976.1 | 1 - 2 | 110 | 118 | ||
GenBank mRNA | BC126068.1 | - | 1 - 2 | 40 | 118 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
