Assay Details
Gene Symbol: Akr1b7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053781.2 | NP_446233.2 | 6 - 7 | 673 | 151 | |
XM_006236250.2 | XP_006236312.1 | 7 - 8 | 1023 | 151 | ||
XM_008762755.1 | XP_008760977.1 | 5 - 6 | 1528 | 151 | ||
XM_008762756.1 | XP_008760978.1 | 5 - 6 | 1528 | 151 | ||
GenBank mRNA | AF182168.1 | - | 6 - 7 | 815 | 151 | |
BC086563.1 | - | 6 - 7 | 673 | 151 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
