Assay Details
Gene Symbol: Bnip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080897.1 | NP_543173.1 | 4 - 5 | 380 | 69 | |
GenBank mRNA | AF454391.1 | - | 4 - 5 | 380 | 69 | |
FQ218385.1 | - | 4 - 5 | 384 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
