Assay Details
Gene Symbol: Fibp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172334.2 | NP_758837.1 | 5 - 6 | 666 | 80 | |
GenBank mRNA | AY093421.1 | - | 5 - 6 | 667 | 80 | |
BC061802.1 | - | 5 - 6 | 687 | 80 | ||
FQ228181.1 | - | 5 - 6 | 718 | 80 | ||
FQ234564.1 | - | 5 - 6 | 718 | 80 | ||
FQ235188.1 | - | 4 - 5 | 643 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
