Assay Details
Gene Symbol: Fibp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172334.2 | NP_758837.1 | 9 - 10 | 1031 | 70 | |
XM_006230701.2 | XP_006230763.1 | 9 - 10 | 1108 | 70 | ||
GenBank mRNA | AY093421.1 | - | 9 - 10 | 1032 | 70 | |
BC061802.1 | - | 9 - 10 | 1052 | 70 | ||
FQ228181.1 | - | 9 - 10 | 1083 | 70 | ||
FQ234469.1 | - | 9 - 10 | 1083 | 70 | ||
FQ234564.1 | - | 9 - 10 | 1083 | 70 | ||
FQ235188.1 | - | 8 - 9 | 1008 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
