Assay Details
Gene Symbol: Tnfaip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182950.4 | NP_891995.1 | 1 - 2 | 167 | 59 | |
GenBank mRNA | AY336949.2 | - | 1 - 2 | 54 | 59 | |
BC128704.1 | - | 1 - 2 | 167 | 59 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AY336949.2, BC128704.1, NM_182950.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
