Assay Details
Gene Symbol: Gabra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183326.2 | NP_899155.1 | 2 - 3 | 301 | 84 | |
XM_006246123.2 | XP_006246185.1 | 2 - 3 | 293 | 84 | ||
XM_006246124.2 | XP_006246186.1 | 2 - 3 | 126 | 84 | ||
GenBank mRNA | AY574250.1 | - | 1 - 2 | 77 | 84 | |
BC100061.1 | - | 2 - 3 | 301 | 84 | ||
FQ211937.1 | - | 2 - 3 | 554 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
