Assay Details
Gene Symbol: Gabra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183326.2 | NP_899155.1 | 3 - 4 | 404 | 85 | |
XM_006246123.2 | XP_006246185.1 | 3 - 4 | 396 | 85 | ||
XM_006246124.2 | XP_006246186.1 | 3 - 4 | 229 | 85 | ||
GenBank mRNA | AY574250.1 | - | 2 - 3 | 180 | 85 | |
BC100061.1 | - | 3 - 4 | 404 | 85 | ||
FQ211937.1 | - | 3 - 4 | 657 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
