Assay Details
Gene Symbol: Gabra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183326.2 | NP_899155.1 | 4 - 5 | 473 | 104 | |
XM_006246123.2 | XP_006246185.1 | 4 - 5 | 465 | 104 | ||
XM_006246124.2 | XP_006246186.1 | 4 - 5 | 298 | 104 | ||
GenBank mRNA | AY574250.1 | - | 3 - 4 | 249 | 104 | |
BC100061.1 | - | 4 - 5 | 473 | 104 | ||
FQ211937.1 | - | 4 - 5 | 726 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
