Assay Details
Gene Symbol: Gabrg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183327.1 | NP_899156.1 | 4 - 5 | 548 | 99 | |
XM_006246125.2 | XP_006246187.1 | 4 - 5 | 1111 | 99 | ||
GenBank mRNA | AY574252.1 | - | 4 - 5 | 548 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
