Assay Details
Gene Symbol: Igsf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198768.2 | NP_942063.1 | 3 - 4 | 849 | 106 | |
GenBank mRNA | AY273816.1 | - | 3 - 4 | 893 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
