Assay Details
Gene Symbol: Pah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012619.2 | NP_036751.2 | 10 - 11 | 1151 | 63 | |
XM_008765215.1 | XP_008763437.1 | 8 - 9 | 1091 | 63 | ||
GenBank mRNA | BC078881.1 | - | 10 - 11 | 1151 | 63 | |
FQ219397.1 | - | 6 - 7 | 587 | 63 | ||
FQ219680.1 | - | 10 - 11 | 1183 | 63 | ||
K02599.1 | - | 5 - 6 | 441 | 63 | ||
M12337.1 | - | 10 - 11 | 1136 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
