Assay Details
Gene Symbol: Angpt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053546.1 | NP_445998.1 | 7 - 8 | 1486 | 77 | |
XM_006241609.2 | XP_006241671.1 | 7 - 8 | 77 | |||
XM_008765463.1 | XP_008763685.1 | 7 - 8 | 77 | |||
XM_008765464.1 | XP_008763686.1 | 7 - 8 | 77 | |||
GenBank mRNA | AB080023.1 | - | 7 - 8 | 1486 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
