Assay Details
Gene Symbol: Tdg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053729.1 | NP_446181.1 | 1 - 2 | 79 | 122 | |
GenBank mRNA | AY026945.1 | - | 1 - 2 | 79 | 122 | |
BC129088.1 | - | 1 - 2 | 96 | 122 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AY026945.1, BC129088.1, NM_053729.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
