Assay Details
Gene Symbol: P4ha3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198775.1 | NP_942070.1 | 11 - 12 | 1496 | 76 | |
XM_008759761.1 | XP_008757983.1 | 12 - 13 | 1574 | 76 | ||
GenBank mRNA | AY313450.1 | - | 11 - 12 | 1496 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
