Assay Details
Gene Symbol: P4ha3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198775.1 | NP_942070.1 | 6 - 7 | 959 | 94 | |
XM_008759761.1 | XP_008757983.1 | 7 - 8 | 1037 | 94 | ||
XM_008759762.1 | XP_008757984.1 | 6 - 7 | 962 | 94 | ||
GenBank mRNA | AY313450.1 | - | 6 - 7 | 959 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
