Assay Details
Gene Symbol: Lcmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199405.2 | NP_955437.1 | 6 - 7 | 681 | 106 | |
XM_006230191.2 | XP_006230253.1 | 6 - 7 | 717 | 106 | ||
GenBank mRNA | BC063186.1 | - | 6 - 7 | 684 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
