Assay Details
Gene Symbol: Frzb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100527.1 | NP_001093997.1 | 2 - 3 | 859 | 63 | |
XM_006234429.2 | XP_006234491.1 | 1 - 2 | 1283 | 63 | ||
GenBank mRNA | BC166428.1 | - | 2 - 3 | 859 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
