Assay Details
Gene Symbol: Frzb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100527.1 | NP_001093997.1 | 3 - 4 | 905 | 66 | |
XM_006234429.2 | XP_006234491.1 | 2 - 3 | 1329 | 66 | ||
GenBank mRNA | BC166428.1 | - | 3 - 4 | 905 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
