Assay Details
Gene Symbol: Csrnp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271225.1 | NP_001258154.1 | 2 - 3 | 228 | 120 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001271225.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
