Assay Details
Gene Symbol: Pced1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012348.1 | NP_001012348.1 | 3 - 4 | 478 | 79 | |
XM_006235015.2 | XP_006235077.1 | 5 - 6 | 953 | 79 | ||
XM_006235016.2 | XP_006235078.1 | 6 - 7 | 2325 | 79 | ||
XM_006235017.2 | XP_006235079.1 | 5 - 6 | 860 | 79 | ||
GenBank mRNA | BC089908.1 | - | 3 - 4 | 478 | 79 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
