Assay Details
Gene Symbol: Pced1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012348.1 | NP_001012348.1 | 4 - 5 | 733 | 89 | |
XM_006235015.2 | XP_006235077.1 | 6 - 7 | 1208 | 89 | ||
XM_006235016.2 | XP_006235078.1 | 7 - 8 | 2580 | 89 | ||
XM_006235017.2 | XP_006235079.1 | 6 - 7 | 1115 | 89 | ||
GenBank mRNA | BC089908.1 | - | 4 - 5 | 733 | 89 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
