Assay Details
Gene Symbol: Apip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106492.1 | NP_001099962.1 | 5 - 6 | 569 | 139 | |
XM_006234638.2 | XP_006234700.1 | 5 - 6 | 618 | 139 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
