Assay Details
Gene Symbol: Pld4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126288.1 | NP_001119760.1 | 5 - 6 | 683 | 88 | |
GenBank mRNA | BC161906.1 | - | 5 - 6 | 591 | 88 | |
FQ230808.1 | - | 475 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
