Assay Details
Gene Symbol: Pld4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126288.1 | NP_001119760.1 | 8 - 9 | 1150 | 64 | |
GenBank mRNA | BC161906.1 | - | 8 - 9 | 1058 | 64 | |
FQ230808.1 | - | 942 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
