Assay Details
Gene Symbol: Pot1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024322.1 | NP_001019493.1 | 10 - 11 | 1286 | 124 | |
XM_006236160.2 | XP_006236222.1 | 11 - 12 | 1378 | 124 | ||
XM_006236161.2 | XP_006236223.1 | 12 - 13 | 1600 | 124 | ||
XM_008762809.1 | XP_008761031.1 | 12 - 13 | 1782 | 124 | ||
XM_008762810.1 | XP_008761032.1 | 12 - 13 | 1727 | 124 | ||
XM_008762811.1 | XP_008761033.1 | 10 - 11 | 1331 | 124 | ||
GenBank mRNA | BC085834.1 | - | 10 - 11 | 1286 | 124 | |
BC090024.1 | - | 5 - 6 | 702 | 124 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
