Assay Details
Gene Symbol: Pot1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024322.1 | NP_001019493.1 | 7 - 8 | 987 | 67 | |
XM_006236160.2 | XP_006236222.1 | 8 - 9 | 1079 | 67 | ||
XM_006236161.2 | XP_006236223.1 | 9 - 10 | 1301 | 67 | ||
XM_008762809.1 | XP_008761031.1 | 9 - 10 | 1483 | 67 | ||
XM_008762810.1 | XP_008761032.1 | 9 - 10 | 1428 | 67 | ||
XM_008762811.1 | XP_008761033.1 | 7 - 8 | 1032 | 67 | ||
GenBank mRNA | BC085834.1 | - | 7 - 8 | 987 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
