Assay Details
Gene Symbol: Bmpr1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024259.1 | NP_001019430.1 | 3 - 4 | 273 | 91 | |
XM_006233400.2 | XP_006233462.1 | 7 - 8 | 1117 | 91 | ||
XM_006233401.2 | XP_006233463.1 | 6 - 7 | 778 | 91 | ||
XM_008761513.1 | XP_008759735.1 | 6 - 7 | 977 | 91 | ||
XM_008761514.1 | XP_008759736.1 | 9 - 10 | 1499 | 91 | ||
GenBank mRNA | BC092609.1 | - | 3 - 4 | 273 | 91 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC092609.1, NM_001024259.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
