Assay Details
Gene Symbol: Akap8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053855.1 | NP_446307.1 | 5 - 6 | 880 | 60 | |
XM_006241047.2 | XP_006241109.1 | 5 - 6 | 1102 | 60 | ||
GenBank mRNA | BC166580.1 | - | 5 - 6 | 880 | 60 | |
U01914.1 | - | 5 - 6 | 855 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
