Assay Details
Gene Symbol: Pex26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106618.2 | NP_001100088.1 | 1 - 2 | 528 | 93 | |
XM_006237235.2 | XP_006237297.1 | 1 - 2 | 1154 | 93 | ||
GenBank mRNA | BC166463.1 | - | 1 - 2 | 528 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
