Assay Details
Gene Symbol: Pex26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106618.2 | NP_001100088.1 | 2 - 3 | 670 | 69 | |
XM_006237235.2 | XP_006237297.1 | 2 - 3 | 1296 | 69 | ||
GenBank mRNA | BC166463.1 | - | 2 - 3 | 670 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
