Assay Details
Gene Symbol: Mrps35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106628.1 | NP_001100098.1 | 7 - 8 | 718 | 106 | |
XM_006237676.1 | XP_006237738.1 | 7 - 8 | 723 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P627330 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 258244 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 914120 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 195323 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379336 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 648862 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 780298 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529626 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622469 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 810679 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 940933 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422282 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 728634 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 51294 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 769273 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58102 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 25644 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 463646 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 350725 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63863 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310967 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 231769 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382564 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 233196 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 281930 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 896885 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409449 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 52075 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150741 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6281 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 60616 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P627330 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P627330 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P627330 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P627330 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 7149 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 2967 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10859731 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10859732 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10859730 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10859727 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10859733 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10859728 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10859726 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10859729 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10859725 | - | - |
Rat Genome 230 2.0 Array | 1372839_at | - | - |
Rat Genome 230 2.0 Array | 1372839_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1372839_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1372839_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0400014916.rn.1 | TC0400002072.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0400014912.rn.1 | TC0400002072.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0400014915.rn.1 | TC0400002072.rn.1 | RTA TC |
Rat Expression Set 230 | 1372839_at | - | - |
Rat Expression Set 230 | 1372839_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10859725 | - | - |
Rat Genome U34 Set | rc_AI045908_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20815446 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1350095 | - | - |
