Assay Details
Gene Symbol: Terf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012464.1 | NP_001012482.1 | 3 - 4 | 498 | 108 | |
GenBank mRNA | BC089888.1 | - | 3 - 4 | 498 | 108 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
