Assay Details
Gene Symbol: Zmpste24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107974.1 | NP_001101444.1 | 5 - 6 | 947 | 118 | |
GenBank mRNA | FQ211274.1 | - | 688 | 118 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
