Assay Details
Gene Symbol: Zmpste24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107974.1 | NP_001101444.1 | 9 - 10 | 1522 | 90 | |
GenBank mRNA | FQ211274.1 | - | 1263 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P047562 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 481995 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 477153 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912143 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 358188 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 451159 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 880162 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 797639 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 626380 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304064 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 719223 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 835231 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786974 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 349021 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 846085 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 147519 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240536 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 532203 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 507506 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 875364 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 662070 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 87797 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588214 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 446625 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 135582 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 164350 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 50498 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 86510 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75236 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P047562 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P047562 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P047562 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P466951 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P506267 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 52524 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10879469 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10879477 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10879471 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10879468 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10879472 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10879473 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10879470 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10879475 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10879474 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10879476 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10879467 | - | - |
Rat Genome 230 2.0 Array | 1399085_PM_at | - | - |
Rat Genome 230 2.0 Array | 1399085_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0500033253.rn.1 | TC0500004267.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500033252.rn.1 | TC0500004267.rn.1 | RTA TC |
Rat Expression Set 230 | 1399085_at | - | - |
Rat Expression Set 230 | 1399085_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10879467 | - | - |
Rat Genome U34 Set | rc_AI175477_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21076024 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1358859 | - | - |
