Assay Details
Gene Symbol: Dhrs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037199.3 | NP_001032276.3 | 3 - 4 | 755 | 69 | |
GenBank mRNA | BC107450.1 | - | 3 - 4 | 755 | 69 | |
EF125189.1 | - | 3 - 4 | 1201 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
