Assay Details
Gene Symbol: Yars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025696.1 | NP_001020867.2 | 10 - 11 | 1293 | 77 | |
GenBank mRNA | BC098795.1 | - | 10 - 11 | 1293 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
