Assay Details
Gene Symbol: Fktn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108667.1 | NP_001102137.1 | 4 - 5 | 702 | 69 | |
XM_006238166.2 | XP_006238228.1 | 6 - 7 | 809 | 69 | ||
XM_006238168.2 | XP_006238230.1 | 5 - 6 | 736 | 69 | ||
XM_006238169.1 | XP_006238231.1 | 4 - 5 | 619 | 69 | ||
XM_008763712.1 | XP_008761934.1 | 5 - 6 | 829 | 69 | ||
XM_008763713.1 | XP_008761935.1 | 6 - 7 | 803 | 69 | ||
XM_008763714.1 | XP_008761936.1 | 5 - 6 | 737 | 69 | ||
XM_008763715.1 | XP_008761937.1 | 5 - 6 | 736 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
