Assay Details
Gene Symbol: Fktn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108667.1 | NP_001102137.1 | 5 - 6 | 977 | 76 | |
XM_006238166.2 | XP_006238228.1 | 7 - 8 | 1084 | 76 | ||
XM_006238168.2 | XP_006238230.1 | 6 - 7 | 1011 | 76 | ||
XM_006238169.1 | XP_006238231.1 | 5 - 6 | 894 | 76 | ||
XM_008763712.1 | XP_008761934.1 | 6 - 7 | 1104 | 76 | ||
XM_008763713.1 | XP_008761935.1 | 7 - 8 | 1078 | 76 | ||
XM_008763714.1 | XP_008761936.1 | 6 - 7 | 1012 | 76 | ||
XM_008763715.1 | XP_008761937.1 | 6 - 7 | 1011 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
