Assay Details
Gene Symbol: Fktn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108667.1 | NP_001102137.1 | 9 - 10 | 1493 | 136 | |
XM_006238166.2 | XP_006238228.1 | 11 - 12 | 1600 | 136 | ||
XM_006238168.2 | XP_006238230.1 | 10 - 11 | 1527 | 136 | ||
XM_006238169.1 | XP_006238231.1 | 9 - 10 | 1410 | 136 | ||
XM_008763712.1 | XP_008761934.1 | 10 - 11 | 1620 | 136 | ||
XM_008763713.1 | XP_008761935.1 | 11 - 12 | 1594 | 136 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P792220 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P196984 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617904 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 111890 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 351678 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 946243 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 254223 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 950755 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 462875 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 170941 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172375 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690086 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 797777 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908101 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 773969 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 676575 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 33663 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 761390 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 242043 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793913 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 568287 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112693 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732187 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 149602 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 652433 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 584911 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 408450 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 8534 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144099 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 139966 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 390824 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 688337 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908220 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2212 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488685 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 201170 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 398052 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 340240 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 918819 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 955041 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 145334 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 165720 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75870 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150696 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 129729 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 67015 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 15105 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 134406 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P196984 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P792220 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P792220 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P196984 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P792220 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P196984 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P196984 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 49791 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10869147 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10869142 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10869144 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10869139 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10869140 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10869148 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10869141 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10869143 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10869145 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10869146 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10869138 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10869137 | - | - |
Rat Genome 230 2.0 Array | 1375785_at | - | - |
Rat Genome 230 2.0 Array | 1375872_PM_at | - | - |
Rat Genome 230 2.0 Array | 1375872_at | - | - |
Rat Genome 230 2.0 Array | 1375785_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1375785_at | - | - |
Affymetrix HT Rat Focus Array | 1375872_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1375872_at | - | - |
Affymetrix HT Rat Focus Array | 1375785_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0500004544.rn.1 | TC0500000663.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500004546.rn.1 | TC0500000663.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500004545.rn.1 | TC0500000663.rn.1 | RTA TC |
Rat Expression Set 230 | 1375785_at | - | - |
Rat Expression Set 230 | 1375785_PM_at | - | - |
Rat Expression Set 230 | 1375872_at | - | - |
Rat Expression Set 230 | 1375872_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10869137 | - | - |
Rat Genome U34 Set | rc_AI144944_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21357634 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1359585 | - | - |
