Assay Details
Gene Symbol: Rad23b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025275.1 | NP_001020446.1 | 7 - 8 | 1110 | 72 | |
GenBank mRNA | BC070960.1 | - | 3 - 4 | 329 | 72 | |
BC090351.1 | - | 4 - 5 | 573 | 72 | ||
BC098674.1 | - | 7 - 8 | 1110 | 72 | ||
BC111406.1 | - | 2 - 3 | 249 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
