Assay Details
Gene Symbol: Nfatc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107805.1 | NP_001101275.1 | 8 - 9 | 2126 | 57 | |
XM_008762507.1 | XP_008760729.1 | 8 - 9 | 2120 | 57 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription by RNA polymerase II
cytokine production
transcription by RNA polymerase II
cellular response to DNA damage stimulus
biological_process
response to xenobiotic stimulus
positive regulation of gene expression
myotube cell development
cell migration
positive regulation of B cell proliferation
calcineurin-NFAT signaling cascade
positive regulation of transcription, DNA-templated
positive regulation of transcription by RNA polymerase II
B cell receptor signaling pathway
positive regulation of myoblast fusion
transcription cis-regulatory region binding
RNA polymerase II cis-regulatory region sequence-specific DNA binding
molecular_function
chromatin binding
DNA-binding transcription factor activity
transcription factor binding
sequence-specific DNA binding
negative regulation of transcription by RNA polymerase II
cytokine production
transcription by RNA polymerase II
cellular response to DNA damage stimulus
biological_process
response to xenobiotic stimulus
positive regulation of gene expression
myotube cell development
cell migration
positive regulation of B cell proliferation
calcineurin-NFAT signaling cascade
positive regulation of transcription, DNA-templated
positive regulation of transcription by RNA polymerase II
B cell receptor signaling pathway
positive regulation of myoblast fusion
transcription cis-regulatory region binding
RNA polymerase II cis-regulatory region sequence-specific DNA binding
molecular_function
chromatin binding
DNA-binding transcription factor activity
transcription factor binding
sequence-specific DNA binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P160499 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 130480 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 15748 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 486957 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 126426 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 533666 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496042 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 493569 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 361084 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 590288 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 361149 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853929 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 72326 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 682925 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 119587 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 755245 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865744 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 316958 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 498645 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 533032 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310100 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 591865 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 765179 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370102 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 680887 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 755816 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 361578 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 361769 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138303 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736924 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 374414 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 957694 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 782422 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382511 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383194 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 777087 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 59133 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158228 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 841313 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 430457 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 299476 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383564 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 670366 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 64602 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858530 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 848135 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 666603 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 762451 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 540392 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 18861 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 343006 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 820155 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 446924 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 104598 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 268557 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 440740 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684323 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 614353 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916717 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 200779 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 451469 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 849608 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 937781 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115192 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 216317 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379562 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 859595 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952871 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 581425 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284965 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372372 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409294 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 917258 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 29742 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 364586 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 424130 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 833407 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876643 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 348715 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10432 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 437916 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115055 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 196058 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 530168 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 42559 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 168005 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 113241 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 72258 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 51492 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 95645 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 119032 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 37951 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 59923 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17055 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 76319 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 27747 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 84482 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 38531 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 124314 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 22719 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P160499 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P160499 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P160499 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P328547 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 64892 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 65156 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10852033 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10852030 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10852025 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10852029 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10852023 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10852031 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10852024 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10852026 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10852021 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10852028 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10852032 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10852019 | - | - |
Affymetrix Rat Clariom S Assay | PSR0300037206.rn.1 | TC0300004852.rn.1 | RTA TC |
Affymetrix Rat Gene 1.0 ST Array | 10852019 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20897829 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1366684 | - | - |
