Assay Details
Gene Symbol: Tubd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105826.1 | NP_001099296.1 | 7 - 8 | 1254 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
